Fmoc-Arg(Pbf)-OH ≥99.0% (by HPLC), powder, Novabiochem®

Supplier: Novabiochem (Part of Merck)

8.52067.0100 8.52067.0025 8.52067.0005 8.52067.0500 8.52067.0250 8.52067.2000 8.52067.5000
04-12-1145-100EA 704 EUR
04-12-1145-100 04-12-1145-25 04-12-1145-5 8.52067.0500 8.52067.0250 NOVA8.52067.2000 NOVA8.52067.5000
Fmoc-Arg(Pbf)-OH ≥99.0% (by HPLC), powder, Novabiochem®
Fmoc-Arg(Pbf)-OH

It can be used for TLC-Systems.

Formula: C₃₄H₄₀N₄O₇S
Melting Pt: > 65 °C
MDL Number: MFCD00235804
CAS Number: 154445-77-9

Order Now

Specification Test Results

Alternate Names N-α-Fmoc-NG-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine
Colour (Visual) White to Off-White
Appearance of Substance (Visual) Powder
Colour Index (0,5 M in DMF) ≤ 60 Hazen
Identity (IR) Passes Test
Enantiomeric Purity ≥ 99,8% (a/a)
Purity (HPLC) ≥ 99,0% (a/a)
Fmoc-ß-Ala-OH (HPLC) ≤ 0,1% (a/a)
Fmoc-ß-Ala-Arg(Pbf)-OH (HPLC) ≤ 0,1% (a/a)
Fmoc-Arg(Pbf)-Arg(Pbf)-OH (HPLC) ≤ 0,1% (a/a)
Fmoc-Arg-OH (HPLC) ≤ 0,1% (a/a)
Assay Free Amino acid (HPLC) ≤ 0,2%
Purity (TLC(011C)) ≥ 98%
Purity (TLC(157B)) ≥ 98%
Solubility (12,5 mmol in 25 ml DMF) Clearly Soluble
Assay (Acidimetric) ≥ 90,0%
Water (K, F,) ≤ 2,0%
Ethyl acetate (HS-GC) ≤ 1,0%
Acetate (IC) ≤ 0,02%
Storage Store at -15 to -25 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR