Heavy duty orbital shakers, analogue
Supplier: OHAUS
30391802 30391837
OHAU30391802EA 2220 EUR
OHAU30391802 OHAU30391837
Heavy duty orbital shakers, analogue
Shakers Orbital Shakers
These heavy duty analogue orbital shakers are designed for a wide range of applications that require basic shaking control. Shakers provide reproducible motion that is evenly distributed throughout the entire surface of the tray. Ideal for bacterial suspensions, staining/destaining, and general mixing.
  • Continuous or timed operation
  • Basic speed and time knobs with dial settings from 1 to 10
  • Speed ramping feature slowly increases speed to desired set-point to avoid splashing
  • Spill-resistant design ensures fluids are channelled away from internal components
More Product Information