Density meter, DMA500
Supplier: Anton Paar
94680
PAAR94680EA 14090 EUR
PAAR94680
Density meter, DMA500
Density Meters
DMA 500 is a laboratory density meter providing measurement with quality.
  • Clear sample identification for full traceability of results
  • Automatic viscosity correction of values
  • Peristaltic pump for error-free filling
  • Integrated rechargeable battery safeguarding the instrument from voltage fluctuations and power outages
  • Easy concentration measurement via preinstalled conversion tables or input of customer specific calculations
FillingCheck™ of this unit automatically detects filling errors and gas bubbles in the filled sample and generates a warningMore Product Information