Nitric acid 2%, Optima® for calibration blank in ICP

Supplier: PERKIN ELMER GC INSTRUMENTS
Warning

N0773120
PERCN0773120EA 173 EUR
PERCN0773120
Nitric acid 2%, Optima® for calibration blank in ICP
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 3264
ADR: 8 III
Merck Index: 13,06610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR