Ruthenium standard solution, 1,000 mg/L Ru in 10% hydrochloric acid (from RuCl3.H2O) standard for AAS and ICP
Supplier: PERKIN ELMER GC INSTRUMENTS
N9303795 N9300146
PERCN9303795EA 231 EUR
PERCN9303795 PERCN9300146
Ruthenium standard solution, 1,000 mg/L Ru in 10% hydrochloric acid (from RuCl3.H2O) standard for AAS and ICP
Standards Cationic Standards Ruthenium Standards