Capillary columns, Elite-200 for autosystem
Supplier: PERKIN ELMER GC INSTRUMENTS
N9316619 N9316621 N9316624 N9316626 N9316628 N9316630 N9316631 N9316633 N9316642 N9316648
PERCN9316619EA 1800 EUR
PERCN9316619 PERCN9316621 PERCN9316624 PERCN9316626 PERCN9316628 PERCN9316630 PERCN9316631 PERCN9316633 PERCN9316642 PERCN9316648
Capillary columns, Elite-200 for autosystem
Chromatography Columns
Elite-200 capillary columns are specially designed for the analysis of solvents.