Cooled Incubators, ST 1
Supplier: POL-EKO APARATURA SP.
ST1CSMART ST1CSSMART ST1PSMART ST1PSSMART ST1PSMARTPRO ST1PSSMARTPRO
POLEST1CSMARTEA 1680 EUR
POLEST1CSMART POLEST1CSSMART POLEST1PSMART POLEST1PSSMART POLEST1PSMARTPRO POLEST1PSSMARTPRO
Cooled Incubators, ST 1
Incubators Cooled Incubators
ST 1 Cooled Incubators ensure reliable temperature control from 3 to 70 °C, regardless of ambient conditions. POL-EKO's models boast internal LED lighting, secure lockable doors. Ideal for stability testing and pharmaceutical storage, these incubators offer versatile chamber configurations, optimising laboratory space and enabling independent temperature control in each compartment.
  • Quality control protocol (at 37 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (according to DIN 12880) for C (comfort)
More Product Information