Ultra-Low Freezers, ZLN-UT 300 VIP
Supplier: POL-EKO APARATURA SP.
ZLNUT300VIPCSM ZLNUT300VIPPSP ZLNUT300VIPPSM
POLEZLNUT300VIPCSMEA 9180 EUR
POLEZLNUT300VIPCSM POLEZLNUT300VIPPSP POLEZLNUT300VIPPSM
Ultra-Low Freezers, ZLN-UT 300 VIP
Freezers Ultra Low Temperature Freezers
Revolutionise your freezing experience with ZLN-UT Ultra-Low Freezers, reaching an impressive ‒86 °C. Manufactured with VIP insulated panels, these freezers ensure long-term preservation for biotechnological samples. These 'As Smart As Your Smartphone' ultra-low freezers offer reliability and flexibility. Choice of either COMFORT model with a stainless steel interior, which is perfect for general use or the PREMIUM model, featuring an acid-resistant stainless steel interior, which is ideal for specialised applications.
  • Quality control protocol (at ‒80 °C)
  • Instruction manual in English
More Product Information