Anti-ZBTB9 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
29-156
PRSI29-156EA 660 EUR
PRSI29-156
Anti-ZBTB9 Rabbit Polyclonal Antibody
Antibodies
ZBTB9 contains 1 BTB (POZ) domain and 2 C2H2-type zinc fingers. It may be involved in transcriptional regulation.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ZBTB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR