Anti-IDH3A Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
29-722
PRSI29-722EA 601 EUR
PRSI29-722
Anti-IDH3A Rabbit Polyclonal Antibody
Antibodies
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD (+) as the electron acceptor and the other NADP (+). Five isocitrate dehydrogenases have been reported: three NAD (+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP (+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD (+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step ofMore Product Information