Anti-PGR Mouse Monoclonal Antibody [clone: PR501]
Supplier: ProSci Inc.
33-190
PRSI33-190EA 773 EUR
PRSI33-190
Anti-PGR Mouse Monoclonal Antibody [clone: PR501]
Antibodies
This antibody is specific to progesterone receptor and shows minimal cross-reaction with other members of the family. Progesterone receptor is expressed as two major isoforms, PR-A (81 kDa) and PR-B (116 kDa). Expression of progesterone receptor has been suggested to reflect a intact estrogen regulatory machinery and therefore predict better clinical response to endocrine therapy than ER alone.
WB: 0,5 - 1 µg/ml Immunohistochemistry: 0,5 - 1 µg/ml (Cat No. V2224 is recommended for IHC) The concentration stated for each application is a general starting point. Variations in protocols,More Product Information