Anti-CLU Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
36-299
PRSI36-299EA 509 EUR
PRSI36-299
Anti-CLU Rabbit Polyclonal Antibody
Antibodies
Clusterin shares homology with the small heat shock protein family of molecular chaperones. The mature secreted form of the protein is a glycosylated, 80-kDa disulfide-linked heterodimer of alpha and beta subunits (produced by internal cleavage). Clusterin is expressed in virtually all tissues and found in all human fluids. It is involved in numerous physiological processes important for carcinogenesis and tumor growth, including apoptotic cell death, cell cycle regulation, DNA repair, cell adhesion, tissue remodeling, lipid transportation, membrane recycling, and immune system regulation.More Product Information