Anti-COL7A1 Goat Polyclonal Antibody

Supplier: ProSci Inc.
46-647
PRSI46-647EA 620 EUR
PRSI46-647
Anti-COL7A1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 250 kDa band observed in rat testis lysates (calculated MW of 159 kDa according to rat NP_001100328.1). Recommended concentration: 0.1-1.5 μg/mL.

Collagen VII antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Collagen VII antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: COL7A1
Clonality: Polyclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR