Anti-GAR1 Chicken Polyclonal Antibody
Supplier: ProSci Inc.
XW-7880
PRSIXW-7880EA 826 EUR
PRSIXW-7880
Anti-GAR1 Chicken Polyclonal Antibody
Antibodies
FUNCTION: Required for ribosome biogenesis and telomere maintenance. Part of the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex, which catalyzes pseudouridylation of rRNA. This involves the isomerization of uridine such that the ribose is subsequently attached to C5, instead of the normal N1. Each rRNA can contain up to 100 pseudouridine ("psi") residues, which may serve to stabilize the conformation of rRNAs. May also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.