Safety boots, pull-on, Steelite™ WORK Rigger FW13
Supplier: Portwest Clothing
FW13BKR48 FW13BKR47 FW13BKR45 FW13BKR43 FW13BKR42 FW13BKR41 FW13BKR40 FW13BKR39 FW13BKR38 FW13BKR44 FW13TAR37 FW13TAR38 FW13TAR39 FW13TAR40 FW13TAR41 FW13TAR42 FW13TAR43 FW13TAR44 FW13TAR45 FW13TAR46 FW13TAR47 FW13TAR48
PRTWFW13BKR48EA 72.8 EUR
PRTWFW13BKR48 PRTWFW13BKR47 PRTWFW13BKR45 PRTWFW13BKR43 PRTWFW13BKR42 PRTWFW13BKR41 PRTWFW13BKR40 PRTWFW13BKR39 PRTWFW13BKR38 PRTWFW13BKR44 PRTWFW13TAR37 PRTWFW13TAR38 PRTWFW13TAR39 PRTWFW13TAR40 PRTWFW13TAR41 PRTWFW13TAR42 PRTWFW13TAR43 PRTWFW13TAR44 PRTWFW13TAR45 PRTWFW13TAR46 PRTWFW13TAR47 PRTWFW13TAR48
Safety boots, pull-on, Steelite™ WORK Rigger FW13
Shoes and Boots
Anti-static pull-on boots made from split buffalo leather and a dual-density PU/rubber, sole resistant to oil.
  • Fur lining
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
  • Side tabs for easy donning
Colour: Black/brown
Certifications: EN ISO 20345:2011; S1P, SRC.