Anti-Rabbit Immunoglobulin Mouse Monoclonal Antibody

Supplier: QED Bioscience
14602
QEDB14602EA 405 EUR
QEDB14602
Anti-Rabbit Immunoglobulin Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Rabbit Immunoglobulin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR