Anti-N-Acetylprocainamide Mouse Monoclonal Antibody

Supplier: QED Bioscience
16908
QEDB16908EA 405 EUR
QEDB16908
Anti-N-Acetylprocainamide Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: N-Acetylprocainamide
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR