Vibratory feeder DR 100 series
Supplier: Retsch GmbH
03.729.0040 03.729.0036 02.785.0019 03.729.0035 03.729.0037 03.729.0039 03.729.0054 03.729.0055 03.785.0159 03.785.0146 02.266.0259 02.266.0260 72.020.0015 72.020.0012 72.020.0011 72.020.0010 72.020.0009 70.938.0001 72.020.0016
RETS03.729.0040EA 206 EUR
RETS03.729.0040 03.729.0036 02.785.0019 03.729.0035 03.729.0037 RETS03.729.0039 RETS03.729.0054 RETS03.729.0055 RETS03.785.0159 RETS03.785.0146 RETS02.266.0259 RETS02.266.0260 RETS72.020.0015 RETS72.020.0012 RETS72.020.0011 RETS72.020.0010 RETS72.020.0009 RETS70.938.0001 RETS72.020.0016
Vibratory feeder DR 100 series
Sample Dividers
The Vibratory feeder is used for uniform continuous feed of pourable bulk materials and fine powders.
  • Reproducible results from uniform feed
  • Digital time setting or continous operation
  • Infinitely adjustable flow volume 0 to 5 litres/minute
  • Easy to set up and operate
  • Different sizes of feed shutes and hoppers to suit a a wide range of materials