Bottles, Wide Neck, without Screw Cap
Supplier: RIXIUS
10300005032OSV 10300012540OSV 10300015045OSV 10300100068OSV 10301003032OSV 10301015045OSV 10301050055ADO 1-0301-0100-45 10301005032OSV 10300003032OSV 10300010040OSV 10300025055OSV 10300050055OSV 10301007540OSV 1-0300-0050-40 10301010040ADO 10301025055ADO 1030010045OSV 1-0301-0500-60 10301005040OSV 10301030055OSV 10301012540OSV 10301020055OSV 10301250080OSV 10300020055OSV 10301100068OSV
RIXI10300005032OSVEA 293 EUR
RIXI10300005032OSV RIXI10300012540OSV RIXI10300015045OSV RIXI10300100068OSV RIXI10301003032OSV RIXI10301015045OSV RIXI10301050055ADO RIXI1-0301-0100-45 RIXI10301005032OSV RIXI10300003032OSV RIXI10300010040OSV RIXI10300025055OSV RIXI10300050055OSV RIXI10301007540OSV 215-4292 RIXI10301010040ADO RIXI10301025055ADO RIXI1030010045OSV RIXI1-0301-0500-60 RIXI10301005040OSV RIXI10301030055OSV 215-4662 215-4663 215-4664 RIXI10300020055OSV RIXI10301100068OSV
Bottles, Wide Neck, without Screw Cap
Bottles Glass Bottles
Clear or amber glass with thread.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes.
Ordering information: Screw caps must be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable