Celite® 503, filter aid

Supplier: ROTH CARL
Warning

0007.1
ROTH0007.1EA 82.5 EUR
ROTH0007.1
Celite® 503, filter aid
Celite® 503, filter aid
Formula: O₂Si
MW: 60.08 g/mol
CAS Number: 68855-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR