Potassium-tert-butoxide

Supplier: ROTH CARL
Danger

3956.1
ROTH3956.1EA 0 EUR
ROTH3956.1
Potassium-tert-butoxide
Potassium-tert-butoxide
Formula: C₄H₉KO
MW: 112.21 g/mol
Melting Pt: 256…258 °C
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 3206
ADR: 4.2 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR