(4-Nitrophenyl)-β-D-glucopyranoside

Supplier: ROTH CARL

4082.1
ROTH4082.1EA 0 EUR
ROTH4082.1
(4-Nitrophenyl)-β-D-glucopyranoside
(4-Nitrophenyl) β-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
MDL Number: MFCD00006593
CAS Number: 2492-87-7
EINECS: 219-661-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR