D-(+)-Mannose

Supplier: ROTH CARL

4220.1 4220.2
ROTH4220.1EA 0 EUR
ROTH4220.1 ROTH4220.2
D-(+)-Mannose
D(+)-Mannose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 376
Melting Pt: 133
Density: 1.539 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064122
CAS Number: 3458-28-4
EINECS: 222-392-4
Merck Index: 13,05772

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR