Methyl butyrate

Supplier: ROTH CARL

5186.1
ROTH5186.1EA 0 EUR
ROTH5186.1
Methyl butyrate
Methyl butyrate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 102
Melting Pt: –84 °C
Density: 0.8984 g/cm³ (20 °C)
Flash Pt: 14
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3 II
Merck Index: 13,06062

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR