L(-)-Glucose

Supplier: ROTH CARL

7509.1
ROTH7509.1EA 0 EUR
ROTH7509.1
L(-)-Glucose
L(-)-Glucose
Formula: C₆H₁₂O₆
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 146…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00148913
CAS Number: 921-60-8
EINECS: 213-068-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR