Lactose monohydrate

Supplier: ROTH CARL

8921.5
ROTH8921.5EA 0 EUR
ROTH8921.5
Lactose monohydrate
Lactose monohydrate
Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Melting Pt: 223
Storage Temperature: Ambient
CAS Number: 10039-26-6
EINECS: 200-559-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR