Ethanol absolute

Supplier: ROTH CARL
Danger

9065.1 9065.2 9065.3 9065.4 9065.6
ROTH9065.1EA 0 EUR
ROTH9065.1 ROTH9065.2 ROTH9065.3 ROTH9065.4 ROTH9065.6
Ethanol absolute
Ethanol
Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78.3
Melting Pt: –117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR