Aluminium potassium sulphate dodecahydrate

Supplier: ROTH CARL

CN78.1
ROTHCN78.1EA 31.7 EUR
ROTHCN78.1
Aluminium potassium sulphate dodecahydrate
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
Melting Pt: 92,5 °C
Density: 1,72 g/cm³ (20 °C)
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR