Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)

Supplier: ROTH CARL

K055.1
ROTHK055.1EA 100 EUR
ROTHK055.1
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Formula: C₉H₁₂N₂Na₃O₁₅P₃
MW: 550.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00044310
CAS Number: 19817-92-6
EINECS: 243-347-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR