Barium hydroxide octahydrate

Supplier: ROTH CARL
Danger

P009.1 P009.2
ROTHP009.1EA 77.3 EUR
ROTHP009.1 ROTHP009.2
Barium hydroxide octahydrate
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Boiling Pt: 780
Melting Pt: 78
Density: 2.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8 III
Merck Index: 13,00980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR