Magnesium acetate tetrahydrate

Supplier: ROTH CARL

P026.1 P026.2
ROTHP026.1EA 82.5 EUR
ROTHP026.1 ROTHP026.2
Magnesium acetate tetrahydrate
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR