Blotting membranes, Amersham™ Hybond™ ECL™
Supplier: Cytiva
RPN82D RPN132D
RPN82DEA 299 EUR
RPN82D RPN132D
Blotting membranes, Amersham™ Hybond™ ECL™
Transfer Membranes
Hybond™ ECL™ gives excellent sensitivity, resolution, and low background for all labelling and detection systems (radioactive, non radioactive, and chromogenic), especially when used with ECL™, ECL™ Prime, and ECL™ Plex Western Blotting Detection kits and reagents. Special packaging features include interleaf on both sides of the membrane and air-tight resealable aluminium bag with label space for marking the 'opened on' date.
  • 100% pure nitrocellulose membrane validated for use with all ECL™ Western blotting systems and for all protein blotting
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable