Blood collecting systems, S-Monovette® EDTA
Supplier: SARSTEDT
03.1068.001 05.1167.001 04.1914.001 04.1901.001 02.1066.001 04.1931.001 01.1605.001 04.1931 05.1167 06.1664.001 02.1333.001 03.1068 04.1901 04.1914 04.1932.001
SARS03.1068.001EA 211 EUR
SARS03.1068.001 SARS05.1167.001 SARS04.1914.001 SARS04.1901.001 SARS02.1066.001 SARS04.1931.001 SARS01.1605.001 SARS04.1931 SARS05.1167 SARS06.1664.001 SARS02.1333.001 SARS03.1068 SARS04.1901 SARS04.1914 SARS04.1932.001
Blood collecting systems, S-Monovette® EDTA
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
S-Monovette® EDTA and needle ensures reliability and safety to blood collection system.
  • Recessed membrane protection against risk of exposure
  • Screw cap to minimise aerosal effect
  • Transparent plastic label