Laboratory bottle, glass, amber, protect, plastic safety coated, narrow neck, GL thread, UV protected, DURAN®
Supplier: DWK Life Sciences
218062438 218063631 218066337 218065435 218061733 218064439 1173548 218061433
SCOT218062438EA 47.8 EUR
SCOT218062438 SCOT218063631 SCOT218066337 SCOT218065435 SCOT218061733 SCOT218064439 SCOT1173548 SCOT218061433
Laboratory bottle, glass, amber, protect, plastic safety coated, narrow neck, GL thread, UV protected, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, protect, amber, without screw cap and pouring ring.
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Purely external application of the colour ensures unchanged DURAN® properties on the interior
  • Glass Type I according to USP, EP and JP
  • Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
Round, with graduation and DIN thread. The coating provides scratch, leak and splinter protection and is ideallyMore Product Information