Adapters, receiver, bent, DURAN®
Supplier: DWK Life Sciences
243100808
SCOT243100808EA 30.7 EUR
SCOT243100808
Adapters, receiver, bent, DURAN®
Distillation Components Distillation Adapters
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bent, with ground socket.