PEEK tubing
Supplier: SGE Analytical Science
1302015008-10F 1301005001-5F 1301007002-5F 1302020004-10F 1302030005-10F 1302030001-10F 1301010003-5F 1301015008-5F 1301020004-5F 1302004006-5F 1302005001-5F 1302007002-5F 1302010003-5F 1302013004-5F 1302015008-5F 1302020004-5F 1302030005-5F 1302030001-5F 1302040000-5F 1303062000-5F 1303080000-5F 1304002000-5F 1304003006-5F 1304004001-5F 1304005005-5F 1304006002-5F 1301005001-10F 1301007002-10F 1301010003-10F 1301015008-10F 1301020004-10F 1302004006-10F 1302005001-10F 1302007002-10F 1302010003-10F 1302013004-10F 1302040000-10F 1303062000-10F 1303080000-10F 1304002000-10F 1304003006-10F 1304004001-10F 1304005005-10F 1304006002-10F 1301005001-50F 1301007002-50F 1301010003-50F 1301015008-50F 1301020004-50F 1302004006-50F 1302005001-50F 1302007002-50F 1302010003-50F 1302013004-50F 1302015008-50F 1302020004-50F 1302030005-50F 1302030001-50F 1302040000-50F 1303062000-50F 1303080000-50F 1304002000-50F 1304003006-50F 1304004001-50F 1304005005-50F 1304006002-50F 1301005001100F 1301007002100F 1301015008100F 1301020004100F 1302004006100F 1302005001100F 1302007002100F 1302010003100F 1302013004100F 1302015008100F 1302020004100F 1302030005100F 1302030001100F 1302040000100F 1303062000100F 1303080000100F 1304002000100F 1304003006100F 1304004001100F 1304005005100F 1304006002100F
SGEA1302015008-10FEA 53.2 EUR
SGEA1302015008-10F SGEA1301005001-5F SGEA1301007002-5F SGEA1302020004-10F SGEA1302030005-10F SGEA1302030001-10F SGEA1301010003-5F SGEA1301015008-5F SGEA1301020004-5F SGEA1302004006-5F SGEA1302005001-5F SGEA1302007002-5F SGEA1302010003-5F SGEA1302013004-5F SGEA1302015008-5F SGEA1302020004-5F SGEA1302030005-5F SGEA1302030001-5F SGEA1302040000-5F SGEA1303062000-5F SGEA1303080000-5F SGEA1304002000-5F SGEA1304003006-5F SGEA1304004001-5F SGEA1304005005-5F SGEA1304006002-5F SGEA1301005001-10F SGEA1301007002-10F SGEA1301010003-10F SGEA1301015008-10F SGEA1301020004-10F SGEA1302004006-10F SGEA1302005001-10F SGEA1302007002-10F SGEA1302010003-10F SGEA1302013004-10F SGEA1302040000-10F SGEA1303062000-10F SGEA1303080000-10F SGEA1304002000-10F SGEA1304003006-10F SGEA1304004001-10F SGEA1304005005-10F SGEA1304006002-10F SGEA1301005001-50F SGEA1301007002-50F SGEA1301010003-50F SGEA1301015008-50F SGEA1301020004-50F SGEA1302004006-50F SGEA1302005001-50F SGEA1302007002-50F SGEA1302010003-50F SGEA1302013004-50F SGEA1302015008-50F SGEA1302020004-50F SGEA1302030005-50F SGEA1302030001-50F SGEA1302040000-50F SGEA1303062000-50F SGEA1303080000-50F SGEA1304002000-50F SGEA1304003006-50F SGEA1304004001-50F SGEA1304005005-50F SGEA1304006002-50F SGEA1301005001100F SGEA1301007002100F SGEA1301015008100F SGEA1301020004100F SGEA1302004006100F SGEA1302005001100F SGEA1302007002100F SGEA1302010003100F SGEA1302013004100F SGEA1302015008100F SGEA1302020004100F SGEA1302030005100F SGEA1302030001100F SGEA1302040000100F SGEA1303062000100F SGEA1303080000100F SGEA1304002000100F SGEA1304003006100F SGEA1304004001100F SGEA1304005005100F SGEA1304006002100F
PEEK tubing
Tubing Chromatography Tubing
Precision bore PEEK tubing is designed specifically for modern LC, LCMS and automation usage.
  • Chemical resistant and inert to most commonly used solvents
  • Bio-inert and bio-compatible
  • Colour coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with polymer and stainless steel fittings
  • Maximum operating temperature of 100 °C
PEEK tubing is a flexible alternative to stainless steel tubing in high pressure applications. Also ensures reproducibility in chromatographic flowMore Product Information