Defined fetal bovine Serum (FBS), HyClone™, U.S. origin
Supplier: GE Healthcare - Hyclone
SH30070.03IH25 SH30070.03IH30 SH30070.03IR25 SH30070.03IR30
SH30070.03IH25EA 1280 EUR
SH30070.03IH25 SH30070.03IH30 SH30070.03IR25 SH30070.03IR30
Defined fetal bovine Serum (FBS), HyClone™, U.S. origin
Sera (Serum)
Highest quality FBS, HyClone US-origin defined FBS is 40 nm-filtered, virus-tested to 9 CFR standard, and supplied with an extensive biochemical profile.
  • Well-defined: assay results for over 50 components, with additional testing available
  • US origin: sourced and processed in the United States with complete traceability to origin
  • Ultra-low endotoxin: ≤10 EU/ml endotoxin for reduced research and production risk
  • Sterile-filtered: serial 40 nm filter process removes up to 8 logs viral challenge
  • No mycoplasma is present
γ-irradiated and heat-inactivatedMore Product Information