Digital microscope
Supplier: TQC
LD6182
SIMELD6182EA 258 EUR
SIMELD6182
Digital microscope
Microscopes
This USB microscope can be connected to a PC in order to determine the structure of surfaces or other objects. It is designed to analyse coating failures, imperfections or treatment quality.
  • USB camera with the optical accuracy of a LED microscope
  • Can be used vertically on ferrous material thanks to three strong magnets
  • Integrated white LEDs adjustable in strength for clear view without reflection
Suitable for measuring, editing and making videos, as well as determining the length, width, height and angle to the objects.
Delivery information: More Product Information