Immersion flow cup viscometers
Supplier: TQC
VF2215 VF2217 VF2216 VF2213
SIMEVF2215EA 257.77 EUR
SIMEVF2215 SIMEVF2217 SIMEVF2216 SIMEVF2213
Immersion flow cup viscometers
Viscometers Flow Cup Viscometers
Dip cups can be used to provide a quick `break-thread' flow time comparative measurements.
  • Unique serial number
  • Titanium anodised aluminium with fixed stainless steel nozzle and handle
  • Long loop handle to allow the cup to be dipped by hand into a liquid container
Suitable for use on site in a manufacturing environment.
Flow times 25 to 150 seconds.
Certifications: DIN 53211
Ordering information: Stand to be ordered separately.