Biopsy processing/embedding cassettes, Micromesh™, M407
Supplier: Simport Scientific
M407-10 M407-11 M407-12 M407-2 M407-3 M407-4 M407-5 M407-6 M407-7 M407-8 M407-9
SIMPM407-10EA 647 EUR
SIMPM407-10 SIMPM407-11 SIMPM407-12 SIMPM407-2 SIMPM407-3 SIMPM407-4 SIMPM407-5 SIMPM407-6 SIMPM407-7 SIMPM407-8 SIMPM407-9
Biopsy processing/embedding cassettes, Micromesh™, M407
Embedding Cassettes Biopsy Processing Cassettes
These cassettes with lids are made of a high-density acetal polymer and keep specimens safely submerged in liquid.
  • One large 27 mm compartment
  • 1676 square opening (0,36 mm), no biopsy pads needed
  • Recessed cover, a great space saving feature
  • Easy-to-remove pre-attached disposable cover
  • Compatible with Primera printers
The one-piece integral lid eliminates the need for separate steel lids. It can be opened and closed as often as necessary and will relock securely without danger of specimen loss. Large labeling areas are located on three sides of theMore Product Information