myo-Inositol ≥97.0% FCC

Supplier: SPECTRUM CHEMICAL MFG

I1004-500GM I1004-2.5KG I1004-12KG
SPCMI1004-500GMEA 725 EUR
SPCMI1004-500GM SPCMI1004-2.5KG SPCMI1004-12KG
myo-Inositol ≥97.0% FCC
myo-Inositol

Inositol, FCC is a vitamin-like substance, sometimes known colloquially as 'vitamin B-8'. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications.


Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₁₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8

Order Now

Specification Test Results

Assay (Dried Basis) 0.97
Melting Range 224 - 227 °C
Calcium (Ca) To Pass Test
Chloride (Cl) max. 0.005 %
Lead (Pb) max. 4 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR