3-Mercapto-1,2-propanediol NF

Supplier: SPECTRUM CHEMICAL MFG

M4080-100ML M4080-500ML M4080-50LBBL
SPCMM4080-100MLEA 415 EUR
SPCMM4080-100ML SPCMM4080-500ML SPCMM4080-50LBBL
3-Mercapto-1,2-propanediol NF
3-Mercapto-1,2-propanediol

Monothioglycerol, NF is a buffering agent excipient. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₃H₈O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00004879
CAS Number: 96-27-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

ASSAY (ANHYDROUS BASIS) 97.0 - 101.0 %
SPECIFIC GRAVITY 1.241 - 1.250
REFRACTIVE INDEX 1.521 - 1.526
pH OF A 1 IN 10 SOLUTION 3.5 - 7.0
RESIDUE ON IGNITION Max. 0.1 %
SELENIUM Max. 30 µg/g
ELEMENTAL IMPURITIES AS REPORTED
WATER DETERMINATION Max. 5.0 %
IDENTIFICATION TO PASS TEST
RESIDUAL SOLVENTS AS REPORTED


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR