L(+)-Serine USP

Supplier: SPECTRUM CHEMICAL MFG

S1023-1KG S1023-25KGBL S1023-12KGBL
SPCMS1023-1KGEA 3170 EUR
SPCMS1023-1KG SPCMS1023-25KGBL SPCMS1023-12KGBL
L(+)-Serine USP
L(+)-Serine

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259
Melting Pt: 215…225 °C
Density: 1.6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR