Saccharin sodium dihydrate, powder USP

Supplier: SPECTRUM CHEMICAL MFG

SO200-12KG SO200-2.5KG
SPCMSO200-12KGEA 0 EUR
SPCMSO200-12KG SPCMSO200-2.5KG
Saccharin sodium dihydrate, powder USP
Saccharin sodium dihydrate

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: C₇H₅NNaO₃S·2H₂O
MW: 242.21 g/mol
MDL Number: MFCD00151213
CAS Number: 6155-57-3
EINECS: 204-886-1

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR