Zinc sulphate heptahydrate ≥99%, granular USP

Supplier: SPECTRUM CHEMICAL MFG
Danger

ZI120-500GM ZI120-12KG ZI120-2.5KG
SPCMZI120-500GMEA 120 EUR
SPCMZI120-500GM SPCMZI120-12KG SPCMZI120-2.5KG
Zinc sulphate heptahydrate ≥99%, granular USP
Zinc sulphate heptahydrate

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: ZnSO₄·7H₂O
Melting Pt: 100 °C
Density: 1,97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR