IntestiCult™ OGM Human Basal Medium
Supplier: STEMCELL TECHNOLOGIES
100-0190
STMC100-0190EA 709 EUR
STMC100-0190
IntestiCult™ OGM Human Basal Medium
Cell Culture Media
Cell culture medium for establishment and maintenance of human colorectal cancer organoids.
Efficiently grow Wnt-independent colorectal tumor organoids using IntestiCult™ OGM Human Basal Medium. Tumoroids have high predictive value as an in vitro system as they retain the characteristics of the tumor of origin. Use IntestiCult™ OGM Human Basal Medium to provide optimal culture conditions for tumor organoid growth and expansion while maintaining tumor heterogeneity. The Wnt-free formulation ensures that Wnt-dependent, normal, healthy intestinal cells do not contaminate the tumoroid culture.
Tumor organoids generated using IntestiCult™ OGM Human Basal Medium are suitable for a variety of applications, including modeling tumor development and screening drugs for efficacy and toxicity. For growing healthy intestinal organoids, please use IntestiCult™ Organoid Growth Medium (Human) or IntestiCult™-SF Organoid Growth Medium (Human). For differentiating human intestinal organoids, refer to IntestiCult™ Organoid Differentiation Medium (Human).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').