mTeSR™ 1 Feeder-Free Maintenance Medium
Supplier: STEMCELL TECHNOLOGIES
85850 85857
STMC85850EA 0 EUR
STMC85850 STMC85857
mTeSR™ 1 Feeder-Free Maintenance Medium
Cell Culture Media
cGMP, feeder-free maintenance medium for human ES and iPS cells.
Use this specialised, feeder-free culture medium to achieve more consistent human pluripotent stem cell (hPSC) cultures with homogenous, undifferentiated phenotypes. Manufactured under relevant cGMPs, mTeSR™1 ensures the highest quality and consistency for reproducible results in your fundamental research, as well as for cell therapy and investigational new drug research applications. This serum-free, complete cell culture medium is made with pre-screened raw materials to ensure batch-to-batch consistency and robust performance in feeder-free hPSC culture.
Use established protocols for applications ranging from derivation to differentiation with this most widely published feeder-free hPSC culture medium, which has been used by leading pluripotent stem cell researchers to successfully maintain thousands of hPSC lines in over 50 countries. For enhanced cell performance and versatile maintenance, you may also be interested in mTeSR™ Plus medium, which is also manufactured under relevant cGMPs and features stabilised components and enhanced buffering.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').