Cole-Parmer® Stuart TR-200 Tube Rotators
Supplier: Cole Parmer
51901-28 51901-26
STUA51901-28EA 912 EUR
STUA51901-28 STUA51901-26
Cole-Parmer® Stuart TR-200 Tube Rotators
Shakers Rotating/Rocking Shakers
Get complete mixing with both rocking and rolling movement.
  • Fully adjustable mixing angle optimizes mixing for any sample
  • 360° rotation
  • Integrated drip tray prevents workspace spills
  • Variety of tube holder discs allows customization
These rotators keep cultures in suspension without compromising. The angle of rotation is fully adjustable with a scale to measure and reproduce the mixing angle, from horizontal for minimal mixing to vertical for full end-over-end mixing. Wide operating conditions allow for safe use in incubators up to 40 °C (generalMore Product Information