Jenway® 76 UV/Visible Scanning Spectrophotometer
Supplier: Cole Parmer
83056-79 83056-82 99959-81
STUA83056-79EA 922 EUR
STUA83056-79 JENW83056-82 STUA99959-81
Jenway® 76 UV/Visible Scanning Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Advanced split beam optics ensure excellent accuracy and reproducibility.
  • Make setup easy with the user-friendly 178 mm high-definition colour touchscreen and interface
  • Choice of measurement modes let you handle a multitude of applications
  • Simple connectivity with multiple USB ports for data storage and printer connectivity
  • Versatile compatible with a wide range of interchangeable accessories
Designed with your convenience in mind, the 76 series of spectrophotometers provide accurate and reliable results in various applications from teaching, toMore Product Information