Retractable safety scalpels
Supplier: SWANN MORTON
3901 3991 4091 4002 4008 4005 4006 3905 4015 4007 3915 3910 4001 3911 4010 4011 3906 3907 3908
SWAN3901EA 125 EUR
SWAN3901 SWAN3991 SWAN4091 SWAN4002 SWAN4008 SWAN4005 SWAN4006 SWAN3905 SWAN4015 SWAN4007 SWAN3915 SWAN3910 SWAN4001 SWAN3911 SWAN4010 SWAN4011 SWAN3906 SWAN3907 SWAN3908
Retractable safety scalpels
Scalpels and Scalpel Assemblies Scalpels
Stainless steel blade, plastic handle. These disposable safety scalpels support and enhance safe passing procedures and protocols already practiced within the operating room and theatres.
  • Colour coded for ease of blade shape recognition
  • Scalpel features a strong temporary forward lock position to prevent uncontrolled retraction when contacting with bone during use
  • Additional notch to hold the blade more securely in the passing position and a rear permanent lock to be engaged prior to final disposal in a sharps container
Packaging: All scalpels areMore Product Information